Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000266742.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000429527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000554226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
NM_001135175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
NM_001135176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
NM_152905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000266742.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000429527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000554226.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
ENST00000557644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
NM_001135175.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
NM_001135176.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
NM_152905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200162699.hg.1 |
TC1200008544.hg.1 |
Clariom D TC |