Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013036.hg.1 |
ENST00000311637.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013036.hg.1 |
ENST00000338821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013036.hg.1 |
ENST00000402822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013036.hg.1 |
NM_006045.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013036.hg.1 |
TC20000944.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013035.hg.1 |
ENST00000311637.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013035.hg.1 |
ENST00000338821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013035.hg.1 |
ENST00000402822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013035.hg.1 |
NM_006045.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013035.hg.1 |
TC20000944.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
ENST00000311637.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
ENST00000338821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
NM_006045.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
TC2000009461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
ENST00000311637.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
ENST00000338821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000166176.hg.1 |
NM_006045.2 |
Refseq |