Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006544.3 | NP_006535.1 | 5 - 6 | 774 | 114 | |
XM_005267272.3 | XP_005267329.1 | 5 - 6 | 859 | 114 | ||
GenBank mRNA | AK295085.1 | - | 5 - 6 | 681 | 114 | |
AK303531.1 | - | 4 - 5 | 517 | 114 | ||
AK312523.1 | - | 5 - 6 | 779 | 114 | ||
BC041126.1 | - | 5 - 6 | 740 | 3914527 | 114 | |
U85946.1 | - | 5 - 6 | 718 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |