Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
TC06001100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
ENST00000253332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
ENST00000354675.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
ENST00000359755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
ENST00000402676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
NM_005100.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013014.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
TC06001100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
ENST00000253332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
ENST00000354675.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
ENST00000359755.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
ENST00000402676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
ENST00000490177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
NM_005100.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013013.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013009.hg.1 |
TC06001100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013009.hg.1 |
ENST00000354675.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013009.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013008.hg.1 |
TC06001100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013008.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167486.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167486.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167486.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167486.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167486.hg.1 |
TC0600009862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000253332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000359755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000402676.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
NM_005100.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000253332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000359755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
ENST00000402676.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
NM_005100.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167491.hg.1 |
TC0600009862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600167487.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167487.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167487.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167487.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167487.hg.1 |
TC0600009862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000253332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000359755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000402676.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000490177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
NM_005100.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000253332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000354675.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000359755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000402676.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
ENST00000490177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
NM_005100.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167490.hg.1 |
TC0600009862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600167485.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167485.hg.1 |
NM_144497.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167485.hg.1 |
TC0600009862.hg.1 |
Clariom D TC |