Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282199.1 | NP_001269128.1 | 3 - 4 | 311 | 146 | |
GenBank mRNA | BI755534.1 | - | 3 - 4 | 287 | 146 | |
D83768.1 | - | 2 - 3 | 161 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |