Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM96A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032231.5 | NP_115607.1 | 3 - 4 | 576 | 87 | |
NR_110310.1 | 4 - 5 | 694 | 87 | |||
GenBank mRNA | AK312171.1 | - | 3 - 4 | 518 | 87 | |
BC008865.2 | - | 3 - 4 | 478 | 4128736 | 87 | |
CN421575.1 | - | 3 - 4 | 504 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |