Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159544.hg.1 |
TC2200008305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200159544.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159544.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159544.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159544.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008305.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159542.hg.1 |
TC2200008305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200159542.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159542.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159542.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159542.hg.1 |
NM_001887.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159545.hg.1 |
TC2200008305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200159545.hg.1 |
ENST00000215939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159545.hg.1 |
ENST00000215939.2 |
Ensembl |