Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014845.5 | NP_055660.1 | 17 - 18 | 2166 | 135 | |
XM_011536281.2 | XP_011534583.1 | 17 - 18 | 2698 | 135 | ||
XM_017011591.1 | XP_016867080.1 | 17 - 18 | 2171 | 135 | ||
XM_017011592.1 | XP_016867081.1 | 16 - 17 | 1934 | 135 | ||
XM_017011593.1 | XP_016867082.1 | 15 - 16 | 1869 | 135 | ||
GenBank mRNA | AK222732.1 | - | 17 - 18 | 135 | ||
AK291671.1 | - | 17 - 18 | 2091 | 135 | ||
BC041338.2 | - | 17 - 18 | 2122 | 5274004 | 135 | |
D87464.1 | - | 17 - 18 | 2075 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |