Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192.2 | NP_001183.2 | 1 - 2 | 352 | 118 | |
GenBank mRNA | BC058291.1 | - | 1 - 2 | 275 | 6736094 | 118 |
EF152353.1 | - | 1 - 2 | 131 | 118 | ||
Z14954.1 | - | 1 - 2 | 200 | 118 | ||
Z29575.1 | - | 1 - 2 | 352 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |