Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029799.hg.1 |
TC01001915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029799.hg.1 |
ENST00000366646.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029799.hg.1 |
ENST00000366647.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029799.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029799.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188240.hg.1 |
TC0100011958.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000366647.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000416000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000436239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000469332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
ENST00000492459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011958.hg.1 |
NM_014236.3 |
Refseq |