Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000465141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000465141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000465141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010037.hg.1 |
ENST00000221459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010037.hg.1 |
ENST00000391864.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010037.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010037.hg.1 |
TC19000725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010038.hg.1 |
ENST00000221459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010038.hg.1 |
ENST00000391864.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010038.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010038.hg.1 |
TC19000725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010041.hg.1 |
ENST00000221459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010041.hg.1 |
ENST00000391864.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010041.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010041.hg.1 |
TC19000725.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
TC1900008526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165673.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000465141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000465141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000465141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000469137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000474252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000486217.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
ENST00000595200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008526.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
NM_022165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
TC1900008526.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
ENST00000221459.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
ENST00000391864.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
NM_001308419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165667.hg.1 |
NM_022165.2 |
Refseq |