Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000468095.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000468095.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000468095.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027198.hg.1 |
TC06002159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027198.hg.1 |
ENST00000343505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027198.hg.1 |
ENST00000427025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027198.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027198.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202704.hg.1 |
TC0600014348.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000342260.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000343505.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000426841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000427025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000468095.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000468095.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000468095.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000479393.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000533765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
ENST00000534376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_001144060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014348.hg.1 |
NM_020464.1 |
Refseq |