Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003656.4 | NP_003647.1 | 3 - 4 | 399 | 69 | |
XM_005265516.1 | XP_005265573.1 | 3 - 4 | 353 | 69 | ||
GenBank mRNA | AB451277.1 | - | 2 - 3 | 214 | 69 | |
AB451407.1 | - | 2 - 3 | 214 | 69 | ||
AK314917.1 | - | 3 - 4 | 353 | 69 | ||
BC106754.2 | - | 3 - 4 | 360 | 40024362 | 69 | |
BC106755.2 | - | 3 - 4 | 360 | 40024363 | 69 | |
BI546222.1 | - | 3 - 4 | 397 | 69 | ||
DN990042.1 | - | 3 - 4 | 412 | 69 | ||
L41816.1 | - | 3 - 4 | 361 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons |