Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
ENST00000301260.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
ENST00000357694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
ENST00000398448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
ENST00000450849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
ENST00000539301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001251.hg.1 |
TC19000078.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000601323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
ENST00000601323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000595916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000598136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000601323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
ENST00000601323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006660.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
NM_033064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
TC1900006660.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
ENST00000450849.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
ENST00000597739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
ENST00000600960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146627.hg.1 |
NM_033064.4 |
Refseq |