Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056954.hg.1 |
TC01003709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056954.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056954.hg.1 |
ENST00000367261.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056954.hg.1 |
ENST00000435533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056954.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228534.hg.1 |
TC0100016969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
ENST00000367261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303051.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_001303109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016969.hg.1 |
NM_021633.3 |
Refseq |