Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015667.hg.1 |
TC09001223.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015667.hg.1 |
ENST00000376736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015667.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015670.hg.1 |
TC09001223.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015670.hg.1 |
ENST00000376736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015670.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015668.hg.1 |
TC09001223.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015668.hg.1 |
ENST00000376736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015668.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171581.hg.1 |
TC0900010462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000479197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010462.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171584.hg.1 |
TC0900010462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171580.hg.1 |
TC0900010462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000376736.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000472900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000476087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000483155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
ENST00000490113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
NM_018339.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171583.hg.1 |
TC0900010462.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000475791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000485282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000490856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012186.hg.1 |
ENST00000592441.1 |
Ensembl |