Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005095.2 | NP_005086.2 | 23 - 24 | 3608 | 130 | |
XM_005271328.4 | XP_005271385.1 | 23 - 24 | 3887 | 130 | ||
XM_005271331.2 | XP_005271388.1 | 20 - 21 | 2853 | 130 | ||
XM_011542421.2 | XP_011540723.1 | 23 - 24 | 3534 | 130 | ||
XM_011542423.2 | XP_011540725.1 | 24 - 25 | 3939 | 130 | ||
XM_011542425.2 | XP_011540727.1 | 21 - 22 | 2893 | 130 | ||
XM_017002803.1 | XP_016858292.1 | 24 - 25 | 3661 | 130 | ||
XM_017002804.1 | XP_016858293.1 | 24 - 25 | 3601 | 130 | ||
XM_017002805.1 | XP_016858294.1 | 19 - 20 | 3099 | 130 | ||
GenBank mRNA | AB007885.1 | - | 19 - 20 | 2728 | 130 | |
AK309705.1 | - | 22 - 23 | 3268 | 130 | ||
BC012093.1 | - | 21 - 22 | 3187 | 130 | ||
BC127113.1 | - | 24 - 25 | 3614 | 40125110 | 130 | |
BC127114.1 | - | 20 - 21 | 2797 | 40125115 | 130 | |
BX647376.1 | - | 19 - 20 | 2839 | 130 | ||
BX648067.1 | - | 14 - 15 | 1902 | 130 | ||
CR936741.1 | - | 22 - 23 | 3472 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |