Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025990.hg.1 |
ENST00000300671.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025990.hg.1 |
ENST00000389840.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025990.hg.1 |
NM_173628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025990.hg.1 |
TC17001918.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000389840.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000585328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000586052.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000590227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000591369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
NM_173628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000389840.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000585328.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000586052.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000590227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
ENST00000591369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
NM_173628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197072.hg.1 |
TC1700011905.hg.1 |
Clariom D TC |