Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002114.3 | NP_002105.3 | 5 - 6 | 6385 | 124 | |
XM_011514546.2 | XP_011512848.1 | 7 - 8 | 7428 | 124 | ||
XM_011514547.2 | XP_011512849.1 | 6 - 7 | 6587 | 124 | ||
XM_011514548.2 | XP_011512850.1 | 6 - 7 | 6831 | 124 | ||
XM_011514549.2 | XP_011512851.1 | 6 - 7 | 6711 | 124 | ||
XM_011514550.2 | XP_011512852.1 | 6 - 7 | 6578 | 124 | ||
XM_011514551.2 | XP_011512853.1 | 6 - 7 | 7256 | 124 | ||
XM_011514552.2 | XP_011512854.1 | 5 - 6 | 6416 | 124 | ||
XM_011514553.1 | XP_011512855.1 | 5 - 6 | 6397 | 124 | ||
XM_011514555.2 | XP_011512857.1 | 4 - 5 | 7986 | 124 | ||
XM_017010800.1 | XP_016866289.1 | 8 - 9 | 7314 | 124 | ||
XM_017010801.1 | XP_016866290.1 | 8 - 9 | 7255 | 124 | ||
XM_017010802.1 | XP_016866291.1 | 6 - 7 | 7334 | 124 | ||
XM_017010803.1 | XP_016866292.1 | 7 - 8 | 7429 | 124 | ||
GenBank mRNA | BC140816.1 | - | 4 - 5 | 6318 | 9021684 | 124 |
M32019.1 | - | 2 - 3 | 3758 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |