Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154606.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154606.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154606.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154606.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154606.hg.1 |
TC1200007655.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000546981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000548970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
ENST00000575924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007655.hg.1 |
NR_120440.1 |
Refseq |