Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
TC03000419.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000314557.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000314589.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000328528.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000352241.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000394351.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000394355.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000448226.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000472437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
ENST00000531774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_000248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_001184967.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_006722.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_198158.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_198159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_198177.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008731.hg.1 |
NM_198178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000314557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000314589.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000328528.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000352241.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000394351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000433517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000448226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000451708.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000461014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000472437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000478490.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000531774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_000248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_001184967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_006722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198158.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198177.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000314557.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000314589.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000328528.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000352241.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000394351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000433517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000448226.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000451708.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000461014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000472437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000478490.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
ENST00000531774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_000248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_001184967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_006722.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198158.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198177.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
NM_198178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300156011.hg.1 |
TC0300007748.hg.1 |
Clariom D TC |