Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001113.hg.1 |
TC01000070.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001113.hg.1 |
ENST00000378344.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001113.hg.1 |
ENST00000456805.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001113.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000344579.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006619.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145774.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145774.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145774.hg.1 |
ENST00000378344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145774.hg.1 |
NM_182752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145774.hg.1 |
TC0100006619.hg.1 |
Clariom D TC |