Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000313581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000411317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000433782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000441800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000461516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000461516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000461516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000464687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000464687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000464687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000464687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000467927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000467927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000467927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000467927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000474303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000474303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000474303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000474303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000474303.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000476208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000476208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000476208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000476208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
ENST00000520309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010253.hg.1 |
NM_144708.3 |
Refseq |