Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000256737.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000525139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000529242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000529242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000529242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000529242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000529242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
ENST00000531646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007106.hg.1 |
NM_031418.3 |
Refseq |