Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153366.3 | NP_699197.3 | 40 - 41 | 10161 | 98 | |
GenBank mRNA | AJ619977.1 | - | 40 - 41 | 9996 | 98 | |
AK075200.1 | - | 3 - 4 | 616 | 98 | ||
AK075234.1 | - | 3 - 4 | 2839 | 98 | ||
AL832416.1 | - | 3 - 4 | 2648 | 98 | ||
AY243503.1 | - | 40 - 41 | 9996 | 98 | ||
AY916667.1 | - | 4 - 5 | 3602 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |