Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082969.1 | NP_001076438.1 | 4 - 5 | 449 | 148 | |
NM_001301019.1 | NP_001287948.1 | 4 - 5 | 490 | 148 | ||
NM_018195.3 | NP_060665.3 | 4 - 5 | 1007 | 148 | ||
NR_103469.1 | 4 - 5 | 1048 | 148 | |||
GenBank mRNA | AB096251.1 | - | 4 - 5 | 353 | 148 | |
BC005403.1 | - | 2 - 3 | 246 | 148 | ||
BC030546.1 | - | 2 - 3 | 251 | 148 | ||
BC048133.1 | - | 4 - 5 | 935 | 5295844 | 148 | |
BG115155.1 | - | 4 - 5 | 497 | 148 | ||
BX538107.1 | - | 4 - 5 | 1006 | 148 | ||
CD244552.1 | - | 4 - 5 | 408 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |