Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002694.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002694.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002694.hg.1 |
TC20000216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148797.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148797.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148797.hg.1 |
TC2000007133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148797.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148797.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |