Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017010900.1 | XP_016866389.1 | 3 - 4 | 447 | 71 | |
GenBank mRNA | LC010245.1 | - | 1 - 2 | 88 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |