Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002609.hg.1 |
ENST00000269491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002609.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002609.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002609.hg.1 |
TC18000227.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002610.hg.1 |
ENST00000269491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002610.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002610.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002610.hg.1 |
TC18000227.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
TC1800007546.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151768.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007546.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
NM_080474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
TC1800007546.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
ENST00000269491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
ENST00000382768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
NM_001307928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151769.hg.1 |
NM_080474.2 |
Refseq |