Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000532281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000532281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
ENST00000532281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000326737.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000529192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000532281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000532281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
ENST00000532281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010710.hg.1 |
NM_173811.3 |
Refseq |