Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197325.1 | NP_001184254.1 | 13 - 14 | 1548 | 100 | |
NM_001286035.1 | NP_001272964.1 | 15 - 16 | 1634 | 100 | ||
NM_001286036.1 | NP_001272965.1 | 14 - 15 | 1593 | 100 | ||
NM_001668.3 | NP_001659.1 | 14 - 15 | 1593 | 100 | ||
NM_178427.2 | NP_848514.1 | 13 - 14 | 1548 | 100 | ||
XM_005245151.1 | XP_005245208.1 | 14 - 15 | 1535 | 100 | ||
XM_005245153.1 | XP_005245210.1 | 14 - 15 | 1520 | 100 | ||
XM_005245154.3 | XP_005245211.1 | 15 - 16 | 1563 | 100 | ||
XM_011509542.2 | XP_011507844.1 | 14 - 15 | 1596 | 100 | ||
XM_011509543.2 | XP_011507845.1 | 15 - 16 | 1591 | 100 | ||
XM_011509544.2 | XP_011507846.1 | 14 - 15 | 1560 | 100 | ||
XM_011509545.2 | XP_011507847.1 | 13 - 14 | 1551 | 100 | ||
XM_011509546.1 | XP_011507848.1 | 13 - 14 | 1311 | 100 | ||
XM_011509547.2 | XP_011507849.1 | 13 - 14 | 1551 | 100 | ||
XM_017001288.1 | XP_016856777.1 | 15 - 16 | 1438 | 100 | ||
XM_017001289.1 | XP_016856778.1 | 15 - 16 | 1423 | 100 | ||
XM_017001290.1 | XP_016856779.1 | 14 - 15 | 1392 | 100 | ||
XM_017001291.1 | XP_016856780.1 | 14 - 15 | 1394 | 100 | ||
GenBank mRNA | AB209877.1 | - | 14 - 15 | 1404 | 100 | |
AK290177.1 | - | 14 - 15 | 1532 | 100 | ||
AK291705.1 | - | 13 - 14 | 1390 | 100 | ||
AK293027.1 | - | 14 - 15 | 1593 | 100 | ||
AK313780.1 | - | 14 - 15 | 1534 | 100 | ||
AK315812.1 | - | 13 - 14 | 1387 | 100 | ||
AL834279.1 | - | 15 - 16 | 1573 | 100 | ||
BC041121.1 | - | 14 - 15 | 1407 | 100 | ||
M69238.1 | - | 14 - 15 | 1448 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |