Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000423939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000423939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000423939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000470921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000470921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
ENST00000470921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012108.hg.1 |
TC02000772.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012108.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012108.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200164199.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200164199.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200164199.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200164199.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200164199.hg.1 |
TC0200009167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000423939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000423939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000423939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000431078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000470921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000470921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
ENST00000470921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009167.hg.1 |
NM_130773.3 |
Refseq |