Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315529.1 | NP_001302458.1 | 24 - 25 | 4846 | 108 | |
NM_006031.5 | NP_006022.3 | 24 - 25 | 4684 | 108 | ||
XM_005261124.4 | XP_005261181.1 | 25 - 26 | 4736 | 108 | ||
XM_011529594.2 | XP_011527896.1 | 25 - 26 | 4784 | 108 | ||
XM_017028362.1 | XP_016883851.1 | 24 - 25 | 4703 | 108 | ||
XM_017028363.1 | XP_016883852.1 | 25 - 26 | 4905 | 108 | ||
GenBank mRNA | AB007862.3 | - | 24 - 25 | 4847 | 108 | |
AF515282.1 | - | 24 - 25 | 4577 | 108 | ||
AK296731.1 | - | 11 - 12 | 2283 | 108 | ||
U52962.1 | - | 25 - 26 | 4596 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |