Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007013.3 | NP_008944.1 | 16 - 17 | 2024 | 68 | |
XM_005250760.3 | XP_005250817.1 | 15 - 16 | 1985 | 68 | ||
XM_005250761.3 | XP_005250818.1 | 16 - 17 | 2065 | 68 | ||
XM_017012993.1 | XP_016868482.1 | 15 - 16 | 1977 | 68 | ||
XM_017012995.1 | XP_016868484.1 | 14 - 15 | 1846 | 68 | ||
XM_017012996.1 | XP_016868485.1 | 13 - 14 | 1764 | 68 | ||
GenBank mRNA | AK292800.1 | - | 16 - 17 | 2039 | 68 | |
AK310683.1 | - | 12 - 13 | 1398 | 68 | ||
AL136739.1 | - | 14 - 15 | 1754 | 68 | ||
AY043361.1 | - | 15 - 16 | 1829 | 68 | ||
AY345857.1 | - | 16 - 17 | 2058 | 68 | ||
BC036065.1 | - | 16 - 17 | 2028 | 5296005 | 68 | |
U96113.1 | - | 9 - 10 | 1178 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |