Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025882.hg.1 |
TC07001676.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025882.hg.1 |
ENST00000275732.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025882.hg.1 |
ENST00000539430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025882.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186324.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186324.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186324.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186324.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186324.hg.1 |
TC0700012013.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000275732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000464111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000471340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
ENST00000472105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012013.hg.1 |
NM_022574.5 |
Refseq |