Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013217.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013217.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013217.hg.1 |
TC14001159.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009256.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
ENST00000247178.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
ENST00000558189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
NM_014924.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400168354.hg.1 |
TC1400009256.hg.1 |
Clariom D TC |