Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000230085.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000349379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000368979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
ENST00000426155.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_001300929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_003795.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012777.hg.1 |
NM_152827.3 |
Refseq |