Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006807.hg.1 |
TC06000593.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006807.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006807.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198776.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198776.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198776.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198776.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198776.hg.1 |
TC0600014129.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
ENST00000244496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014129.hg.1 |
NM_033112.3 |
Refseq |