Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000553926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000553926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002754.hg.1 |
ENST00000298119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002754.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002754.hg.1 |
TC14000256.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000553926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000553926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000554171.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
ENST00000555279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006998.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148664.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148664.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148664.hg.1 |
ENST00000298119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148664.hg.1 |
NM_152447.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148664.hg.1 |
TC1400006998.hg.1 |
Clariom D TC |