Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000426.3 | NP_000417.2 | 18 - 19 | 2637 | 72 | |
NM_001079823.1 | NP_001073291.1 | 18 - 19 | 2637 | 72 | ||
XM_005266981.3 | XP_005267038.1 | 18 - 19 | 2637 | 72 | ||
XM_005266982.3 | XP_005267039.1 | 18 - 19 | 2637 | 72 | ||
XM_011535820.2 | XP_011534122.1 | 18 - 19 | 2637 | 72 | ||
XM_017010851.1 | XP_016866340.1 | 18 - 19 | 5025 | 72 | ||
XM_017010852.1 | XP_016866341.1 | 7 - 8 | 3914 | 72 | ||
XM_017010853.1 | XP_016866342.1 | 18 - 19 | 2637 | 72 | ||
GenBank mRNA | Z26653.1 | - | 18 - 19 | 2581 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |