Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021179.hg.1 |
ENST00000260746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021179.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021179.hg.1 |
TC10001624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180034.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180034.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180034.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180034.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180034.hg.1 |
TC1000011690.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
ENST00000260746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011690.hg.1 |
NM_004311.3 |
Refseq |