Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014716.hg.1 |
ENST00000228850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014716.hg.1 |
ENST00000545990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014716.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014716.hg.1 |
TC12001124.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014718.hg.1 |
ENST00000228850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014718.hg.1 |
ENST00000545990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014718.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014718.hg.1 |
TC12001124.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000544636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000544636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014717.hg.1 |
ENST00000228850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014717.hg.1 |
ENST00000545990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014717.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014717.hg.1 |
TC12001124.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014720.hg.1 |
ENST00000228850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014720.hg.1 |
ENST00000545990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014720.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014720.hg.1 |
TC12001124.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172202.hg.1 |
TC1200009676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172200.hg.1 |
TC1200009676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000544636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000544636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172207.hg.1 |
TC1200009676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172209.hg.1 |
TC1200009676.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000540967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000544636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000544636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009676.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000228850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000536414.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000541484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
ENST00000545990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
NM_001278309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
NM_006422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200172197.hg.1 |
TC1200009676.hg.1 |
Clariom D TC |