Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007124.2 | NP_009055.2 | 45 - 46 | 6685 | 72 | |
XM_005267127.4 | XP_005267184.1 | 45 - 46 | 7020 | 72 | ||
XM_005267130.2 | XP_005267187.1 | 46 - 47 | 6903 | 72 | ||
XM_005267133.2 | XP_005267190.1 | 45 - 46 | 7703 | 72 | ||
XM_011536101.2 | XP_011534403.1 | 47 - 48 | 7364 | 72 | ||
XM_011536102.2 | XP_011534404.1 | 47 - 48 | 6905 | 72 | ||
XM_011536106.2 | XP_011534408.1 | 46 - 47 | 7552 | 72 | ||
XM_011536107.2 | XP_011534409.1 | 46 - 47 | 6928 | 72 | ||
XM_017011243.1 | XP_016866732.1 | 47 - 48 | 6902 | 72 | ||
XM_017011244.1 | XP_016866733.1 | 47 - 48 | 6905 | 72 | ||
XM_017011245.1 | XP_016866734.1 | 47 - 48 | 6905 | 72 | ||
GenBank mRNA | X69086.1 | - | 45 - 46 | 6593 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |