Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC017766.1 | - | 3 | 1401 | 105 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | F-box protein 10 | FBXO10 |
FBX10, PRMT11 |
4.4kb | Sense | Chr. 9 - 37509920 - 37576459 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |