Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000358337.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000504897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000506393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000511232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
ENST00000520764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008864.hg.1 |
NM_017706.4 |
Refseq |