Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012961.hg.1 |
ENST00000284509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012961.hg.1 |
ENST00000559829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012961.hg.1 |
NM_024837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012961.hg.1 |
TC15001347.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000284509.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000557955.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000558906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000559726.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000559829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NM_024837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NR_073596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NR_073597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NR_073598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000284509.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000557955.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000558906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000559726.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
ENST00000559829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NM_024837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NR_073596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NR_073597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
NR_073598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168862.hg.1 |
TC1500009377.hg.1 |
Clariom D TC |