Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100184846.hg.1 |
TC1100011128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000398868.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000535789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000538786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
ENST00000540699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_001290048.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011128.hg.1 |
NM_015459.4 |
Refseq |