Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004065.hg.1 |
ENST00000399799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004065.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004065.hg.1 |
TC18000397.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000578051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000578051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000578051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000582445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000582445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000582445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000583556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000583556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000583556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
TC1800008219.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156122.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000573767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000576266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
ENST00000608049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006435.hg.1 |
NR_033770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000399799.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000578051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000578051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000578051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000582445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000582445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000582445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000583556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000583556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000583556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
ENST00000584875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008219.hg.1 |
NM_005406.2 |
Refseq |