Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000433747.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009064.hg.1 |
ENST00000608916.1 |
Ensembl |