Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008695.hg.1 |
TC04000598.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008695.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008695.hg.1 |
ENST00000433996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008695.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008694.hg.1 |
TC04000598.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008694.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008694.hg.1 |
ENST00000433996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008694.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
ENST00000394488.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008534.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158498.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158498.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158498.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158498.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158498.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400158499.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158499.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158499.hg.1 |
ENST00000296499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158499.hg.1 |
NM_004784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158499.hg.1 |
TC0400008534.hg.1 |
Clariom D TC |